Anti-CXCR3 Rabbit Polyclonal Antibody

Supplier: Prosci
7647
10062-052EA 716.01 CAD
10062-052
Anti-CXCR3 Rabbit Polyclonal Antibody
Antibodies
The CXCR3 protein is a G protein-coupled receptor with selectivity for three chemokines, termed CXCL9/Mig (monokine induced by interferon-gamma), CXCL10/IP10 (interferon-gamma-inducible 10 kDa protein) and CXCL11/I-TAC (interferon-inducible T cell alpha-chemoattractant) . Binding of chemokines to this protein induces cellular responses that are involved in leukocyte traffic, most notably integrin activation, cytoskeletal changes and chemotactic migration. Similar to other chemokine receptors such as CXCR4, CXCR3 has been implicated in several malignancies as a biomarker of tumor behavior as well as a potential therapeutic target.

CXCR3 antibody can be used for detection of CXCR3 by Western blot at 1 - 2 ?g/ml.

CXCR3 antibody is affinity chromatography purified via peptide column.

CXCR3 antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CXCR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR