Anti-PPARGC1A Rabbit Polyclonal Antibody

Supplier: Prosci
7705
10062-586EA 716.01 CAD
10062-586
Anti-PPARGC1A Rabbit Polyclonal Antibody
Antibodies
The peroxisome proliferator-activated receptor gamma coactivator 1-alpha (PPARGC1A), also known as LEM-6, is a transcriptional coactivator that regulates the genes involved in energy metabolism. PPARGC1A interacts with PPARgamma, which permits the interaction of PPARGC1A with multiple transcription factors. PPARGC1A can interact with, and regulate the activities of, cAMP response element binding protein (CREB) and nuclear respiratory factors (NRFs). It provides a direct link between external physiological stimuli and the regulation of mitochondrial biogenesis, and is a major factor that regulates muscle fiber type determination. PPARGC1A may be also involved in the development of obesity.

PPARGC1A antibody can be used for detection of PPARGC1A by Western blot at 1 - 2 ?g/ml. Antibody can also be used for Immunohistochemistry starting at 5 ?g/mL. For immunofluorescence start at 20 ?g/mL.

PPARGC1A antibody is affinity chromatography purified via peptide column.

PPARGC1A antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PPARGC1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR