Anti-STAT4 Rabbit Polyclonal Antibody

Supplier: Prosci
79-124
10070-208EA 730.35 CAD
10070-208
Anti-STAT4 Rabbit Polyclonal Antibody
Antibodies
Carries out a dual function: signal transduction and activation of transcription. Involved in IL12 signaling.

Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100

Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.

Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STAT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR