Order Entry
ContactUsLinkComponent
Anti-KDR Rabbit Polyclonal Antibody
Anti-KDR Rabbit Polyclonal Antibody
Catalog #: 10068-640
Supplier:  Prosci
Anti-KDR Rabbit Polyclonal Antibody
Catalog #: 10068-640
Supplier:  Prosci
Price
864,70 $
Log in for availability
Unit of Measure Each (100µG)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    kinase insert domain receptor (a type III receptor tyrosine kinase)
  • Clonality:
    Polyclonal
  • Gene ID:
    P35968
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Antigen symbol:
    KDR
  • Conjugation:
    Unconjugated
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    KDR
  • Modification:
    Tyr 1059
  • Storage buffer:
    Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
  • Molecular weight:
    230 kDa
  • Storage temperature:
    Store antibody at –20 °C for up to one year.
  • Concentration:
    1 mg/mL
  • Shipping temperature:
    4 °C
  • Immunogen:
    VEGFR2 (phospho-Tyr1059) antibody was raised against a peptide sequence around phosphorylation site of tyrosine 1059 (P-D-Y (p) -V-R) derived from Human VEGFR2.
  • Purification:
    Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.
  • Cat. no.:
    10068-640

Specifications

About this item

Receptor for VEGF or VEGFC. Has a tyrosine-protein kinase activity. The VEGF-kinase ligand/receptor signaling system plays a key role in vascular development and regulation of vascular permeability. In case of HIV-1 infection, the interaction with extracellular viral Tat protein seems to enhance angiogenesis in Kaposi's sarcoma lesions

Western Blot: 1:1000

Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.

Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KDR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Customers Who Bought This Also Bought