Anti-CTNNB1 Rabbit Polyclonal Antibody

Supplier: Prosci
79-544
10070-066EA 718.22 CAD
10070-066
Anti-CTNNB1 Rabbit Polyclonal Antibody
Antibodies
Involved in the regulation of cell adhesion and in signal transduction through the Wnt pathway.

Western Blot: 1:500~1:1000

Antibodies were purified by affinity-chromatography using epitope-specific peptide.

Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CTNNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR