Anti-PRF1 Mouse Monoclonal Antibody [clone: CE2.10]

Supplier: Prosci
XA-1009
10068-292EA 966.3 CAD
10068-292
Anti-PRF1 Mouse Monoclonal Antibody [clone: CE2.10]
Antibodies
Perforin antibody can be used for detection of horse Perforin by western blot. This product also recognizes human, mouse, rat, sheep, goat and monkey apoPerforin. Method: Cell extracts from the T cell clone B6.1 (2x106) were resolved by SDS-PAGE under reducing conditions, transferred to nitrocellulose and probed with the CE2.10 antibody at 1 ?g/mL. Proteins were visualized using a peroxidase-conjugated antibody to mouse IgG (SBA) and a chemiluminescence detection system.

PBS containing 0.02% sodium azide

Perforin antibody can be stored at 4°C, stable for one year.

Type: Primary
Antigen: PRF1
Clonality: Monoclonal
Clone: CE2.10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR