Anti-NLRP3 Mouse Monoclonal Antibody

XA-1032
10070-758EA 737.49 CAD
10070-758
Anti-NLRP3 Mouse Monoclonal Antibody
NALP3 antibody can be used to recognize human NALP3. It detects endogenous protein by Immunocytochemistry (1:100) and Western Blot (1:1,000). Method: Following fixation with paraformaldehyde 3.7%, THP1 cells were incubated with the MAb to NALP3 (human) (Nalpy3-a) at a 1:100 dilution. An anti-mouse IgG antibody conjugated with Alexa Fluor 488 was used for detection. This product is for research use only.

PBS containing 0.02% sodium azide

NALP3 antibody can be stored at 4°C, stable for one year.

Type:
Antigen: NLRP3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR