Anti-GDNF Chicken Polyclonal Antibody

Supplier: Prosci
XG-6107
10068-094EA 639.63 CAD
10068-094
Anti-GDNF Chicken Polyclonal Antibody
Antibodies
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR