Anti-DNAJC28 Chicken Polyclonal Antibody

Supplier: Prosci
XG-6118
10068-114EA 639.63 CAD
10068-114
Anti-DNAJC28 Chicken Polyclonal Antibody
Antibodies
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DNAJC28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR