Anti-KCNJ6 Chicken Polyclonal Antibody

Supplier: Prosci
XG-6166
10068-166EA 639.63 CAD
10068-166
Anti-KCNJ6 Chicken Polyclonal Antibody
Antibodies
This antibody can be used at 1:500 to 1:5,000 for immunoblot testing in combination with Anti_IgY HRP or Anti-Igy AP for Western blot analysis

PBS Buffer containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KCNJ6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR