Anti-GHRL Chicken Polyclonal Antibody

Supplier: Prosci
XG-6170
10068-174EA 909.87 CAD
10068-174
Anti-GHRL Chicken Polyclonal Antibody
Antibodies
This antibody can be used at 1:500 to 1:4,000 as detecting antibody in ELISA testing.

PBS Buffer containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GHRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR