Anti-HSPA13 Chicken Polyclonal Antibody

Supplier: Prosci
XG-6194
10068-196EA 640.47 CAD
10068-196
Anti-HSPA13 Chicken Polyclonal Antibody
Antibodies
This antibody can be used at 1:500 to 1:5,000 in combination with Anti-IgY HRP or Anti-IgY AP for Western blot analysis.

PBS Buffer containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HSPA13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR