Anti-Cntf Rabbit Polyclonal Antibody (Biotin)

Supplier: Prosci
XP-5114BT
10098-410EA 786.51 CAD
10098-410
Anti-Cntf Rabbit Polyclonal Antibody (Biotin)
Antibodies
Ciliary neurotrophic factor (CNTF) is expressed in glial cells within the central and peripheral nervous systems. CNTF stimulates gene expression, cell survival or differentiation in a variety of neuronal cell types such as sensory, sympathetic, ciliary and motor neurons. CNTF itself lacks a classical signal peptide sequence of a secreted protein, but is thought to convey its cytoprotective effects after release from adult glial cells by some mechanism induced by injury.

ELISA: To detect rat CNTF by direct ELISA (using 100 µL/well antibody solution) this antibody can be used at a concentration of 0.15 - 0.30 µg/mL. Used in conjunction with compatible secondary reagents, allows the detection of at least 0.2 ng/well of recombinant rat CNTF. Sandwich: To detect Rat CNTF by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25 - 1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our Polyclonal Anti-Rat CNTF (XP-5114) as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant Rat CNTF. Western Blot: To detect rat CNTF by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 mg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant rat CNTF is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

Anti-rat CNTF specific antibody was purified by affinity chromatography and then biotinylated.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Cntf
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR