Anti-PROK1 Goat Polyclonal Antibody (Biotin)

Supplier: Prosci
XP-5121BT
10098-434EA 786.51 CAD
10098-434
Anti-PROK1 Goat Polyclonal Antibody (Biotin)
Antibodies
ELISA:<br><br>To detect hEG-VEGF by direct ELISA (using 100 ?L/well antibody solution) this antibody can be used at a concentration of 0.15 - 0.30 ?g/mL. Used in conjunction with compatible secondary reagents, allows the detection of at least 0.2 ng/well of recombinant hEG-VEGF. <br><br>Sandwich: To detect hEG-VEGF by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.25 - 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our Polyclonal Anti-Human EG-VEGF (XP-5121) as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hEG-VEGF. <br><br>Western Blot:<br>To detect hEG-VEGF by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hEG-VEGF is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PROK1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR