Anti-CXCL5 Rabbit Polyclonal Antibody (Biotin)

Supplier: Prosci
XP-5123BT
10098-440EA 786.51 CAD
10098-440
Anti-CXCL5 Rabbit Polyclonal Antibody (Biotin)
Antibodies
ENA-78 is a a 9.9kD inflammatory chemokine that belongs to the CXC chemokine family. ENA-78 is expressed by fibroblasts, is induced by bacterial lipopolysaccharides, and is chemotactic for neutrophils. This chemokine is produced concomitantly with Interleukin 8 (IL8) in response to stimulation with either Interleukin 1 (IL1) or Tumor Necrosis Factor alpha (TNFA). This chemokine is a potent chemotaxin involved in neutrophil activation.

ELISA: To detect hENA-78 by direct ELISA (using 100 µL/well antibody solution) this antibody can be used at a concentration of 0.15 - 0.30 µg/mL. Used in conjunction with compatible secondary reagents, allows the detection of at least 0.2 ng/well of recombinant hENA-78. Sandwich: To detect hENA-78 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25 - 1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our Polyclonal Anti-Human ENA-78 (XP-5123) as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hENA-78. Western Blot: To detect hENA-78 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hENA-78 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

Anti-hENA-78 specific antibody was purified by affinity chromatography and then biotinylated.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CXCL5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR