Anti-IL10 Rabbit Polyclonal Antibody (Biotin)

Supplier: Prosci
XP-5161BT
10097-848EA 786.32 CAD
10097-848
Anti-IL10 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Interleukins (ILs) are a large group of cytokines that are produced mainly by leukocytes, although some are produced by certain phagocytes and auxiliary cells. ILs have a variety of functions, but most function to direct other immune cells to divide and differentiate. Each IL acts on a specific, limited group of cells through a receptor specific for that IL. Human IL10 is a non glycosylated polypeptide consisting of 178 amino acids. There is 73% homology between the human and mouse IL10 proteins, however, the human IL10 acts on both human and mouse target cells, while the mouse IL10 has species specific activity. The cellular sources of IL10 are CD4+ T cells and T cell clones, thymocytes, B cells and B cell lymphomas, macrophages, mast cell lines and keratinocytes. IL10 will stimulate the growth of stem cells, mast cells and thymocytes. IL10 enhances cytotoxic T cell development, and costimulates B cell differentiation and immunoglobulin secretion. IL10 inhibits cytokine production by macrophages and suppresses macrophage class II MHC expression. The human IL10 gene is on human chromosome

ELISA: To detect hIL-10 by direct ELISA (using 100 µL/well antibody solution) this antibody can be used at a concentration of 0.15 - 0.30 µg/mL. Used in conjunction with compatible secondary reagents, allows the detection of at least 0.2 ng/well of recombinant hIL-10. Sandwich To detect hIL-10 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25 - 1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our Polyclonal Anti-Human IL-10 (XP-5161) as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hIL-10. Western Blot: To detect hIL-10 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hIL-10 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

Anti-hIL-10 specific antibody was purified by affinity chromatography and then biotinylated.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IL10
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR