Anti-IL1A Rabbit Polyclonal Antibody

Supplier: Prosci
XP-5174
10097-866EA 786.51 CAD
10097-866
Anti-IL1A Rabbit Polyclonal Antibody
Antibodies
Neutralization:<br> To yield one-half maximal inhibition [ND50] of the biological activity of hIL-1? (20 ng/mL), a concentration of 1.0 - 2.0 ?g/mL of this antibody is required. <br><br>ELISA:<br><br>To detect hIL-1? by direct ELISA (using 100 ?L/well antibody solution) a concentration of at least 0.5 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant hIL-1?. <br><br>Sandwich:<br><br>To detect hIL-1? by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our Biotinylated Anti-Human IL-1? (XP-5174Bt) as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hIL-1?.<br><br>Western Blot:<br><br>To detect hIL-1? by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hIL-1? is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IL1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR