Anti-IL4 Rabbit Polyclonal Antibody (Biotin)

Supplier: Prosci
XP-5190BT
10098-136EA 786.51 CAD
10098-136
Anti-IL4 Rabbit Polyclonal Antibody (Biotin)
Antibodies
ELISA:<br><br>by direct ELISA (using 100 ?L/well antibody To detect rat IL-4 solution) this antibody can be used at a concentration of 0.15 - 0.30 ?g/mL. Used in conjunction with compatible secondary reagents, allows the detection of at least 0.2 ng/well of recombinant rat IL-4. <br><br>Sandwich:<br><br>To detect Rat IL-4 by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.25 - 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our Polyclonal Anti-Rat IL-4 (XP-5190) as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant Rat IL-4.<br><br>Western Blot:<br><br>To detect rat IL-4 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant rat IL-4 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Il4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR