Anti-IL7 Rabbit Polyclonal Antibody (Biotin)

Supplier: Prosci
XP-5198BT
10098-150EA 786.51 CAD
10098-150
Anti-IL7 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Iterleukin 7 is a lymphoid cell growth factor that affects pre-B, pro-B, and early T cells. IL-7 was previously known as pre-B cell growth factor and lymphopoietin 1. IL-7 supports the growth of early B cells from long-term lymphoid bone marrow cultures. It is mitogenic to thymocytes and enhances the response of cells to other stimuli such as polyhydroxyalkanoate (PHA) and concanavalin A (ConA). IL-7 stimulates the proliferation of CD4+/CD8+ cells. The proliferative response of thymocytes to IL-7 is not affected by antibodies to the T cell growth factors such as IL-2, IL-4 and IL-6, suggesting that IL-7 is capable of stimulating T cell proliferation through a pathway independent of the known T cell growth factors. Mature T cells respond to IL-7 and Con A, but not to IL-7 alone. In myeloid lineage cells, IL-7 upregulates the production of pro-inflammatory cytokines and stimulates the tumoricidal activity of monocytes/macrophages. IL-7 is expressed by adherent stromal cells from various tissues.

ELISA: To detect mIL-7 by direct ELISA (using 100 µL/well antibody solution) this antibody can be used at a concentration of 0.15 - 0.30 µg/mL. Used in conjunction with compatible secondary reagents, allows the detection of at least 0.2 ng/well of recombinant mIL-7. Sandwich: To detect mIL-7 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25 - 1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our Polyclonal Anti-Murine IL-7 (XP-5198) as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant mIL-7. Western Blot: To detect mIL-7 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant mIL-7 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

Anti-mIL-7 specific antibody was purified by affinity chromatography and then biotinylated.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Il7
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR