Anti-CXCL5 Rabbit Polyclonal Antibody

Supplier: ProSci
XP-5216
10097-690EA 786.51 CAD
10097-690
Anti-CXCL5 Rabbit Polyclonal Antibody
Antibodies
The specific function of this protein remains unknown.

ELISA: To detect mLIX by direct ELISA (using 100 µL/well antibody solution) a concentration of at least 0.5 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant mLIX. Sandwich: To detect mLIX by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5 - 2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our Biotinylated Anti-Murine LIX (XP-5216Bt) as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant mLIX. Western Blot: To detect mLIX by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant mLIX is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

Anti-mLIX specific antibody was purified by affinity chromatography employing immobilized mLIX matrix.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Cxcl5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR