Anti-CCL8 Rabbit Polyclonal Antibody

Supplier: ProSci
XP-5220
10097-700EA 786.32 CAD
10097-700
Anti-CCL8 Rabbit Polyclonal Antibody
Antibodies
MCP 2 is a chemotactic factor that attracts monocytes, lymphocytes, basophils and eosinophils. MCP 2 is important to inflammatory host responses, and is found in the highest concentration in the small intestine and peripheral blood cells.

ELISA: To detect hMCP-2 by direct ELISA (using 100 µL/well antibody solution) a concentration of at least 0.5 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant hMCP-2. Sandwich: To detect hMCP-2 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5 - 2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our Biotinylated Anti-Human MCP-2 (XP-5220Bt) as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hMCP-2. Western Blot: To detect hMCP-2 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hMCP-2 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

Anti-hMCP-2 specific antibody was purified by affinity chromatography employing immobilized hMCP-2 matrix.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CCL8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR