Anti-PPBP Rabbit Polyclonal Antibody (Biotin)

Supplier: ProSci
XP-5244BT
10097-554EA 786.51 CAD
10097-554
Anti-PPBP Rabbit Polyclonal Antibody (Biotin)
Antibodies
NAP2 is a platelet-derived growth factor that belongs to the CXC chemokine family. This growth factor is a potent chemoattractant and activator of neutrophils. It has been shown to stimulate various cellular processes including DNA synthesis, mitosis, glycolysis, intracellular cAMP accumulation, prostaglandin E2 secretion, and sythesis of hyaluronic acid and sulfated glycosaminoglycan. It also stimulates the formation and secretion of plasminogen activator by synovial cells.

ELISA: To detect hNAP-2 by direct ELISA (using 100 µL/well antibody solution) this antibody can be used at a concentration of 0.15 - 0.30 µg/mL. Used in conjunction with compatible secondary reagents, allows the detection of at least 0.2 ng/well of recombinant hNAP-2. Sandwich: To detect hNAP-2 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25 - 1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our Polyclonal Anti-Human NAP-2 (XP-5244) as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hNAP-2. Western Blot: To detect hNAP-2 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hNAP-2 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

Anti-hNAP-2 specific antibody was purified by affinity chromatography and then biotinylated.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PPBP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR