Anti-PDGFB Rabbit Polyclonal Antibody

Supplier: ProSci
XP-5255
10097-766EA 786.51 CAD
10097-766
Anti-PDGFB Rabbit Polyclonal Antibody
Antibodies
Neutralization:<br> To yield one-half maximal inhibition [ND50] of the biological activity of hPDGF-BB (10.0 ng/mL), a concentration of 0.05 - 0.07 ?g/mL of this antibody is required. <br><br>ELISA:<br><br>To detect hPDGF-BB by direct ELISA (using 100 ?L/well antibody solution) a concentration of at least 0.5 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant hPDGF-BB. <br><br>Sandwich:<br><br>To detect hPDGF-BB by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our Biotinylated Anti-Human PDGF-BB (XP-5255Bt) as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hPDGF-BB.<br><br>Western Blot:<br><br>To detect hPDGF-BB by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hPDGF-BB is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PDGFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR