Anti-RETN Goat Polyclonal Antibody (Biotin)

Supplier: ProSci
XP-5260BT
10098-114EA 786.51 CAD
10098-114
Anti-RETN Goat Polyclonal Antibody (Biotin)
Antibodies
Resistin, also known as Adipocyte Secreted Factor (ADSF) and FIZZ3, is a cytokine that is specifically secreted by adipocytes and seems to suppress the ability of insulin to stimulate glucose uptake into adipose cells. While the function of resistin is not well understood, it appears to have key paracrine roles and has a suggested role in type 2 diabetes.

ELISA: To detect mResistin by direct ELISA (using 100 µL/well antibody solution) this antibody can be used at a concentration of 0.15 - 0.30 µg/mL. Used in conjunction with compatible secondary reagents, allows the detection of at least 0.2 ng/well of recombinant mResistin. Sandwich: To detect mResistin by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25 - 1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our Polyclonal Anti-Murine Resistin (XP-5260) as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant mResistin. Western Blot: To detect mResistin by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant mResistin is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

Anti-mResistin specific antibody was purified by affinity chromatography and then biotinylated.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Retn
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR