Anti-CD40L Rabbit Polyclonal Antibody

XP-5262
10098-112EA 600.05 CAD
10098-112
Anti-CD40L Rabbit Polyclonal Antibody
ELISA:<br><br>To detect hsCD40L by direct ELISA (using 100 ?L/well antibody solution) a concentration of at least 0.5 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant hsCD40L.<br><br>Sandwich:<br><br>To detect hsCD40-Ligand by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our Biotinylated Anti-Human sCD40-Ligand (XP-5262Bt) as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hsCD40-Ligand. <br><br>Western Blot:<br>To detect hsCD40L by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hsCD40L is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CD40L
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR