Anti-KCNB2 Rabbit Polyclonal Antibody

Supplier: ProSci
XPS-2027
10075-460EA 1078.7 CAD
10075-460
Anti-KCNB2 Rabbit Polyclonal Antibody
Antibodies
Voltage-gated K+ channels are important determinants of neuronal membrane excitability, and differences in K+ channel expression patterns and densities contribute to the variations in action potential waveforms and repetitive firing patterns evident in different neuronal cell types. The delayed rectifier-type (IK)channels (Kv1.5, Kv2.1, and Kv2.2) are expressed on all neuronal somata and proximal dendrites and are also found in a wide variety on non-neuronal cells types including pancreatic islets, alveolar cells and cardiac myocytes. Kv2.1 and Kv2.2 form distinct populations of K+ channels and these subunits are thought to be primarily responsible for IK in superior cervical ganglion cells.

Potassium Channel antibody can be used for specific immunolabeling of the ~125k Kv 2.2 protein in Western blots. Applications include Dot Blots (DB), Immunohistochemistry (IHC) and Western Blots (WB). Rabbit anti-Potassium Channel, Kv2.2 recognizes Kv2.2 in rat and Xenopus. When internally tested under ideal conditions the working dilutions were 1:1000 for IHC, DB and WB.

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Reconstitute in 50 ?l PBS. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Kcnb2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat, Xenopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR