Anti-AKT2 Chicken Polyclonal Antibody

Supplier: ProSci
XW-7021
10071-770EA 1200.74 CAD
10071-770
Anti-AKT2 Chicken Polyclonal Antibody
Antibodies
Murine thymoma viral (v-akt) homolog-2; rac protein kinase beta. AKT2 is a putative oncogene encoding a protein belonging to a subfamily of serine/threonine kinases containing SH2-like (Src homology 2-like) domains. Furthermore, AKT2 was shown to be amplified and overexpressed in 2 of 8 ovarian carcinoma cell lines and 2 of 15 primary ovarian tumors. Overexpression of AKT2 contributes to the malignant phenotype of a subset of human ductal pancreatic cancers. AKT2 is a general protein kinase capable of phophorylating several known proteins.

AKT2 antibody can be used for the detection of AKT2 by Western blot, ELISA, may also work for IHC and ICC.

Antigen affinity-purified

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AKT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR