Anti-AP2M1 Chicken Polyclonal Antibody

Supplier: ProSci
XW-7030
10071-030EA 1200.38 CAD
10071-030
Anti-AP2M1 Chicken Polyclonal Antibody
Antibodies
Adaptor-related protein complex 2, mu 1 subunit. This protein is a subunit of the heterotetrameric coat assembly protein complex 2 (AP2), which belongs to the adaptor complexes medium subunits family. It is required for the activity of a vacuolar ATPase, which is responsible for proton pumping occurring in the acidification of endosomes and lysosomes. It may also play an important role in regulating the intracellular trafficking and function of CTLA-4 protein.

AP2M1 antibody can be used for the detection of AP2M1 by Western blot, may also work for IHC and ICC.

Antigen affinity-purified

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AP2M1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR