Anti-CAPG Chicken Polyclonal Antibody

Supplier: ProSci
XW-7067
10071-032EA 1269.69 CAD
10071-032
Anti-CAPG Chicken Polyclonal Antibody
Antibodies
Capping protein (actin filament), gelsolin-like. This protein, first purified from rabbit alveolar macrophages, was named macrophage capping protein. The protein was subsequently renamed Cap G (gene symbol CAPG). 'Cap' was chosen because unlike other members of the gelsolin/villin family, this protein caps the barbed ends of actin filaments but does not sever them. 'G' was included in the name because the derived amino acid sequence of Cap G most closely resembles gelsolin, possessing 49% identity with its amino terminal half.

CAPG antibody can be used for the detection of CAPG by Western blot, may also work for IHC and ICC.

Antigen affinity-purified

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CAPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR