Anti-CSRP2 Chicken Polyclonal Antibody

Supplier: ProSci
XW-7127
10071-056EA 1269.69 CAD
10071-056
Anti-CSRP2 Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal IgY Antibodies against Cysteine and glycine-rich protein 2, antigen affinity-purified. Systeine and glycine-rich protein 2 (CSRP2) is a member of the CSRP family of genes, encoding a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. CRP2 contains two copies of the cysteine-rich amino acid sequence motif (LIM) with putative zinc-binding activity, and may be involved in regulating ordered cell growth. Other genes in the family include CSRP1 and CSRP3.

CSRP2 antibody can be used for the detection of CSRP2 by Western blot, may also work for IHC and ICC.

Antigen affinity-purified

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CSRP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR