Anti-DCLRE1C Chicken Polyclonal Antibody

XW-7140
10071-024EA 596.37 CAD
10071-024
Anti-DCLRE1C Chicken Polyclonal Antibody
Artemis; DNA cross-link repair 1C, SCIDA, SNM1C, RS-SCID. Human Artemis, a new identified 78.4 KD, novel DNA double-strand break repair/v(d)j recombination protein, has been linked to human severe combined immune deficiency disease due to Artemis mutations. This gene is located at human chromosome 10.

DCLRE1C antibody can be used for the detection of DCLRE1C by Western blot, may also work for IHC and ICC.

Antigen affinity-purified

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DCLRE1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR