Anti-EGFR Chicken Polyclonal Antibody

Supplier: ProSci
XW-7180
10071-072EA 1200.74 CAD
10071-072
Anti-EGFR Chicken Polyclonal Antibody
Antibodies
Epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian); ERBB. Epidermal growth factor receptor, a 1,210-amino acid sequence of the human epidermal growth factor (EGF) receptor precursor, reveals close similarity between the entire predicted v-erb-B mRNA oncogene product and the receptor transmembrane and cytoplasmic domains.

ERBB antibody can be used for the detection of ERBB by Western blot, may also work for IHC and ICC.

Antigen affinity-purified

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EGFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR