Anti-CK8 Chicken Polyclonal Antibody

XW-7333
10069-786EA 541.35 CAD
10069-786
Anti-CK8 Chicken Polyclonal Antibody
Keratin, type II cytoskeletal 8 (Cytokeratin 8). KRT8 is the type II intermediate filament chain keratin. Keratin 8, together with its filament partner keratin 18, are perhaps the most commonly found members of the intermediate filament gene family. They are expressed in single layer epithelial tissues of the body. Mutations in this gene have been linked to cryptogenic cirrhosis.

KRT8 GTEL antibody can be used for the detection of KRT8 GTEL by Western blot, may also work for IHC and ICC.

Antigen affinity-purified

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CK8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR