Anti-LEP Chicken Polyclonal Antibody

Supplier: ProSci
XW-7340
10071-426EA 1269.69 CAD
10071-426
Anti-LEP Chicken Polyclonal Antibody
Antibodies
Leptin; leptin (murine obesity homolog); obesity. Leptin, a product of the Obese (ob) gene, is a ligand for the OB receptor (OC-R). Mice with mutations of the ob gene have been found to be obese and diabetic and to have reduced activity, metabolism, and body temperature. M.W: 16,000.

Leptin antibody can be used for the detection of Leptin by Western blot, ELISA, may also work for IHC and ICC.

Antigen affinity-purified

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR