Anti-RGS16 Chicken Polyclonal Antibody

Supplier: ProSci
XW-7526
10069-836EA 1269.69 CAD
10069-836
Anti-RGS16 Chicken Polyclonal Antibody
Antibodies
Regulator of G-protein signalling 16 , RGS16. This protein negatively regulates G protein-coupled receptor signalling.

RGS16 antibody can be used for the detection of RGS16 by Western blot, may also work for IHC and ICC.

Antigen affinity-purified

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RGS16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR