Anti-PGR Chicken Polyclonal Antibody

Supplier: ProSci
XW-7678
10071-148EA 1200.74 CAD
10071-148
Anti-PGR Chicken Polyclonal Antibody
Antibodies
FUNCTION: The steroid hormones and their receptors are involved in the regulation of eukaryotic gene expression and affect cellular proliferation and differentiation in target tissues.
SIMILARITY: Contains 1 nuclear receptor DNA-binding domain.
SUBCELLULAR LOCATION: Nuclear.
DOMAIN: Composed of three domains: a modulating N-terminal domain, a DNA-binding domain and a C-terminal steroid-binding domain.

Progesterone receptor antibody can be used for the detection of Progesterone receptor by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PGR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR