Anti-CP Chicken Polyclonal Antibody

Supplier: ProSci
XW-7698
10071-176EA 1200.74 CAD
10071-176
Anti-CP Chicken Polyclonal Antibody
Antibodies
FUNCTION: Ceruloplasmin is a blue, copper-binding (6-7 atoms per molecule) glycoprotein found in plasma. Four possible functions are ferroxidase activity, amine oxidase activity, copper transport and homeostasis, and superoxide dismutase activity.
DISEASE: Ceruloplasmin levels are decreased in Wilson disease, in which copper cannot be incorporated into ceruloplasmin in liver because of defects in the copper-transporting ATPase 2.

Ceruloplasmin antibody can be used for the detection of Ceruloplasmin by ELISA and Western Blot, may also work for ICC and IHC.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR