Anti-CXCL13 Chicken Polyclonal Antibody

Supplier: ProSci
XW-7744
10069-852EA 1269.69 CAD
10069-852
Anti-CXCL13 Chicken Polyclonal Antibody
Antibodies
FUNCTION: Chemotactive for B lymphocytes but not for T-lymphocytes, monocytes and neutrophils. Does not induce calcium release in B lymphocytes. Binds to BLR1/CXCR5.
SIMILARITY: Belongs to the intercrine beta (chemokine CxC) family.
SUBCELLULAR LOCATION: Secreted protein.
TISSUE SPECIFICITY: Highest levels in liver, followed by spleen, lymph node, appendix and stomach. Low levels in salivary gland, mammary gland and fetal spleen.

Small inducible cytokine B13 antibody can be used for the detection of Small inducible cytokine B13 by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CXCL13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR