Anti-PPP2R3C Chicken Polyclonal Antibody

XW-7782
10069-744EA 596.37 CAD
10069-744
Anti-PPP2R3C Chicken Polyclonal Antibody
Hypothetical protein FLJ20644 antibody can be used for the detection of Hypothetical protein FLJ20644 by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PPP2R3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR