Anti-EWSR1 Chicken Polyclonal Antibody

Supplier: ProSci
XW-7783
10071-242EA 1269.69 CAD
10071-242
Anti-EWSR1 Chicken Polyclonal Antibody
Antibodies
This protein is a putative RNA binding protein of unknown function. Mutations in this gene, specifically a t(11;22)(q24;q12) translocation, are known to cause Ewing sarcoma as well as neuroectodermal and various other tumors. Alternative splicing of this gene results in two products.

EWSR1 protein antibody can be used for the detection of EWSR1 protein by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EWSR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR