Anti-GSKIP Chicken Polyclonal Antibody

XW-7787
10069-894EA 596.37 CAD
10069-894
Anti-GSKIP Chicken Polyclonal Antibody
UPF0279 protein C14orf129 antibody can be used for the detection of UPF0279 protein C14orf129 by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GSKIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR