Anti-ITGB8 Chicken Polyclonal Antibody

XW-7802
10069-908EA 541.35 CAD
10069-908
Anti-ITGB8 Chicken Polyclonal Antibody
FUNCTION: Integrin beta-V/beta-8 is a receptor for fibronectin.
SUBUNIT: Heterodimer of an beta and a beta subunit. beta-8 associates with beta-V.
SIMILARITY: Contains 1 VWFA domain.
SUBCELLULAR LOCATION: Membrane; single-pass type I membrane protein.
TISSUE SPECIFICITY: Placenta, kidney, brain, ovary, uterus and in several transformed cells. Transiently expressed in 293 human embryonic kidney cells.

Integrin ?-8 antibody can be used for the detection of Integrin ?-8 by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ITGB8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR