Anti-TNFAIP1 Chicken Polyclonal Antibody

Supplier: ProSci
XW-7928
10071-296EA 1269.69 CAD
10071-296
Anti-TNFAIP1 Chicken Polyclonal Antibody
Antibodies
This protein was identified as a gene whose expression can be induced by the tumor necrosis factor beta (TNF) in umbilical vein endothelial cells. Studies of a similar gene in mouse suggest that the expression of this gene is developmentally regulated in a tissue-specific manner.

Tumor necrosis factor, ?-induced protein 1 antibody can be used for the detection of Tumor necrosis factor, ?-induced protein 1 by ELISA and Western Blot, may also work for ICC and IHC.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TNFAIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR