Anti-PA2G4 Chicken Polyclonal Antibody

Supplier: ProSci
XW-7934
10071-300EA 1269.71 CAD
10071-300
Anti-PA2G4 Chicken Polyclonal Antibody
Antibodies
Proliferation-associated protein 2G4 antibody can be used for the detection of Proliferation-associated protein 2G4 by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PA2G4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR