Anti-VPS28 Chicken Polyclonal Antibody

Supplier: ProSci
XW-7975
10071-330EA 1269.69 CAD
10071-330
Anti-VPS28 Chicken Polyclonal Antibody
Antibodies
VPS28 protein homolog antibody can be used for the detection of VPS28 protein homolog by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: VPS28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR