Anti-ATG5 Chicken Polyclonal Antibody

Supplier: ProSci
XW-7984
10071-648EA 1200.74 CAD
10071-648
Anti-ATG5 Chicken Polyclonal Antibody
Antibodies
FUNCTION: May play an important role in the apoptotic process, possibly within the modified cytoskeleton. Its expression is a relatively late event in the apoptotic process, occurring downstream of caspase activity.PTM: Conjugated to ATG12; which is essential for autophagy, but is not required for association with isolation membrane.
SIMILARITY: Belongs to the ATG5 family.

Autophagy protein 5 antibody can be used for the detection of Autophagy protein 5 by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ATG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR