Anti-IGFBP6 Chicken Polyclonal Antibody

Supplier: ProSci
XW-7995
10069-920EA 1269.69 CAD
10069-920
Anti-IGFBP6 Chicken Polyclonal Antibody
Antibodies
FUNCTION: IGF-binding proteins prolong the half-life of the IGFs and have been shown to either inhibit or stimulate the growth promoting effects of the IGFs on cell culture. They alter the interaction of IGFs with their cell surface receptors.PTM: O-linked glycans consist of hexose (probably Gal), N-acetylhexosamine (probably GalNAc) and sialic acid residues. Major glycoforms consist of 8-16 monosaccharides (by homology to IGFBP-6 expressed recominantly in CHO cells).

Insulin-like growth factor binding protein 6 antibody can be used for the detection of Insulin-like growth factor binding protein 6 by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IGFBP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR