Anti-NHP2L1 Chicken Polyclonal Antibody

Supplier: ProSci
XW-7997
10070-016EA 1269.69 CAD
10070-016
Anti-NHP2L1 Chicken Polyclonal Antibody
Antibodies
FUNCTION: Binds to the 5'stem-loop of U4 snRNA and may play a role in the late stage of spliceosome assembly. The protein undergoes a comformational change upon RNA-binding.
SUBUNIT: Interacts with RAD17.
SIMILARITY: Belongs to the ribosomal protein L7Ae family.
SUBCELLULAR LOCATION: Nuclear; nucleolar. Concentrated in the dense fibrillar component of the nucleolus.

NHP2-like protein 1 antibody can be used for the detection of NHP2-like protein 1 by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NHP2L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR