Anti-PLIN2 Chicken Polyclonal Antibody

XW-8024
10069-958EA 547.83 CAD
10069-958
Anti-PLIN2 Chicken Polyclonal Antibody
Adipophilin antibody can be used for the detection of Adipophilin by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PLIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR