Anti-NAGK Chicken Polyclonal Antibody

Supplier: ProSci
XW-8044
10069-930EA 1269.69 CAD
10069-930
Anti-NAGK Chicken Polyclonal Antibody
Antibodies
FUNCTION: Converts endogenous N-acetylglucosamine (GlcNAc), a major component of complex carbohydrates, from lysosomal degradation or nutritional sources into GlcNAc 6-phosphate. Also has ManNAc kinase activity (By similarity).
CATALYTIC ACTIVITY: ATP + N-acetyl-D-glucosamine = ADP + N-acetyl-D-glucosamine 6-phosphate.
TISSUE SPECIFICITY: Ubiquitous.

N-acetylglucosamine kinase antibody can be used for the detection of N-acetylglucosamine kinase by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NAGK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR